Learn More
Hach Company Rack-Drying Glassware-Large
Supplier:  Hach Company 2089010
Encompass
Vinyl-coated steel-wire drying rack for test tubes vials flasks cylinders beakers and other laboratory glassware Rack can be used standing up lying down or mounted to the wall Incorporated drain pan design keeps your counter dry Choose small capacity rack to hold 26 pieces of glassware or the large capacity rack to hold 90 pieces of glassware Dimensions are 47 x 18 4 x 48 3cm (18 5 x 7 25 x 19 inches) Image shown is representative of smaller drying rack
Catalog No. 50-002-57138