Learn More
Integra Biosciences Corp RACK F/0.5 ML MICROCNTFG TUBES
SDP
Catalog No. NC2035490
Encompass
Rack for 0.5 ml Microcentrifuge Tubes.Only compatible with 4, 6 or 8 channel VOYAGER tip spacing pipettesStores up to 48 0.5 ml microcentrifuge tubesFormat 8x6Individual sliders allows a contamination free loading of the tubes
Catalog No. NC2035490 Supplier Integra Biosciences Corp Supplier No. 4541
Rack for 0.5 ml Microcentrifuge Tubes.Only compatible with 4, 6 or 8 channel VOYAGER tip spacing pipettesStores up to 48 0.5 ml microcentrifuge tubesFormat 8x6Individual sliders allows a contamination free loading of the tubes