Learn More
Thomas Scientific Microcentrifuge Floating Rack, holds 24 tubes
SDP
Catalog No. 501461008
Encompass
Compact and Economical Incubate up to twenty-four 1.5ml microcentrifuge tubes in a water bath. Easy to load Microtubes are held snugly and then released when the rack is pressed down on a benchtop Dimensions: 11.9 x 7.8cm (4-11/16x 3-1/16")
Catalog No. 50-146-1008 Supplier Thomas Scientific Supplier No. 1226C47
Compact and Economical Incubate up to twenty-four 1.5ml microcentrifuge tubes in a water bath. Easy to load Microtubes are held snugly and then released when the rack is pressed down on a benchtop Dimensions: 11.9 x 7.8cm (4-11/16x 3-1/16")