Learn More
Thomas Scientific Floating Test Tube Rack, Circle, for 0.5 and 1.5ml microtubes
SDP
Catalog No. 501461006
Encompass_Preferred
Polyfoam racks for PCR, microtubes and centrifuge tubes Eliminate expensive plastic floating racks with economical polyfoam floating racks. Easy to use in waterbaths or beakers. Inscriptions with Identi-Marker pens may be made for quick identification. Unique Features Economical Easy to use Non-absorbent Sizing Circle - 83 x 9.5mm (3-1/4x 1/8") Octagon - 83 x 6.4mm (3-1/4x 1/4") Triangle - 86 x 13mm (3-3/8x 1/2")
Catalog No. 50-146-1006 Supplier Thomas Scientific Supplier No. 1201U02
Polyfoam racks for PCR, microtubes and centrifuge tubes Eliminate expensive plastic floating racks with economical polyfoam floating racks. Easy to use in waterbaths or beakers. Inscriptions with Identi-Marker pens may be made for quick identification. Unique Features Economical Easy to use Non-absorbent Sizing Circle - 83 x 9.5mm (3-1/4x 1/8") Octagon - 83 x 6.4mm (3-1/4x 1/4") Triangle - 86 x 13mm (3-3/8x 1/2")