Learn More
Thomas Scientific Floating Test Tube Rack, Triangle, for 15 and 50ml centrifuge tubes
SDP
Catalog No. 501461007
Encompass
Polyfoam racks for PCR, microtubes and centrifuge tubes Eliminate expensive plastic floating racks with economical polyfoam floating racks. Easy to use in waterbaths or beakers. Inscriptions with Identi-Marker pens may be made for quick identification. Unique Features Economical Easy to use Non-absorbent Sizing Circle - 83 x 9.5mm (3-1/4x 1/8") Octagon - 83 x 6.4mm (3-1/4x 1/4") Triangle - 86 x 13mm (3-3/8x 1/2")
Catalog No. 50-146-1007 Supplier Thomas Scientific Supplier No. 1201U04
Polyfoam racks for PCR, microtubes and centrifuge tubes Eliminate expensive plastic floating racks with economical polyfoam floating racks. Easy to use in waterbaths or beakers. Inscriptions with Identi-Marker pens may be made for quick identification. Unique Features Economical Easy to use Non-absorbent Sizing Circle - 83 x 9.5mm (3-1/4x 1/8") Octagon - 83 x 6.4mm (3-1/4x 1/4") Triangle - 86 x 13mm (3-3/8x 1/2")