Learn More
BIOSMITH RACK FOR 1.5-2.0ML TUBES
Catalog No. NC1592728
Encompass
Anodized aluminum tube rack holds up to forty-eight (48) indivdual 1.5 - 2.0ml tubes. Can be used in ice tray to keep samples cold while working at lab bench.
Catalog No. NC1592728 Supplier BIOSMITH Supplier No. 81003
Anodized aluminum tube rack holds up to forty-eight (48) indivdual 1.5 - 2.0ml tubes. Can be used in ice tray to keep samples cold while working at lab bench.