Learn More
Electron Microscopy Sciences Rack for 1 dram Shell Vial, 1 each
SDP
Catalog No. 50949308
Encompass_Preferred
This rugged polypropylene rack holds 48 vials, size 1 dram (15x45 mm); or 4 ml tissue culture vials; or 2ml serum vials in 4x12 configuration with alpha-numeric indexing. The rack is constructed from chemically resistant, autoclavable (121 degree C for 20 minutes) polypropylene and features corner posts to assure stability when stacking empty.
Catalog No. 50-949-308 Supplier Electron Microscopy Sciences Supplier No. 7264101
This rugged polypropylene rack holds 48 vials, size 1 dram (15x45 mm); or 4 ml tissue culture vials; or 2ml serum vials in 4x12 configuration with alpha-numeric indexing. The rack is constructed from chemically resistant, autoclavable (121 degree C for 20 minutes) polypropylene and features corner posts to assure stability when stacking empty.