Learn More
Thomas Scientific Pipettor Rack for Gilson Pipetman, for Desktop Storage
SDP
Catalog No. 501460481
Encompass_Preferred
Three Styles to Suit Your Needs Choose from 3 styles of epoxy-coated aluminum rack to hold Gilson Pipetman® pipettors. Provides storage and easy access for up to 10 pipettors plus up to two Pipet-Aid® pipettes 0211J40 (H18962-0001) for desktop storage 1226D30 (H18962-0002) for wall mounting; has two holes for screw attachment to the wall and double faced tape (included) 1226D31 (H18962-0004) has extended base with raised lip to hold plastic tip boxes; suitable for wall mounting or for free standing on a bench
Catalog No. 50-146-0481 Supplier Thomas Scientific Supplier No. 0211J40
Three Styles to Suit Your Needs Choose from 3 styles of epoxy-coated aluminum rack to hold Gilson Pipetman® pipettors. Provides storage and easy access for up to 10 pipettors plus up to two Pipet-Aid® pipettes 0211J40 (H18962-0001) for desktop storage 1226D30 (H18962-0002) for wall mounting; has two holes for screw attachment to the wall and double faced tape (included) 1226D31 (H18962-0004) has extended base with raised lip to hold plastic tip boxes; suitable for wall mounting or for free standing on a bench