Learn More
USA Scientific Inc 12-place (3 x 4) upright freezer rack for 100-place hinged boxes
Catalog No. NC9426351
Encompass
12-place (3 long x 4 high) upright freezer rack for 100-place hinged boxes, 18 3/4 in L x 5 7/8 in W x 9 1/4 H, stainless steel
Catalog No. NC9426351 Supplier USA Scientific Inc Supplier No. 26101200
12-place (3 long x 4 high) upright freezer rack for 100-place hinged boxes, 18 3/4 in L x 5 7/8 in W x 9 1/4 H, stainless steel