Learn More
Thomas Scientific Upright Freezer Rack for 15 and 50 ml Tube Boxes, 2 x 2, Capacity 4 boxes 12 3/16 x 10 3/16 x 5 7/8" (L x H x W)
SDP
Catalog No. 501459967
Encompass_Preferred
Denville freezer boxes for 15ml and 50ml centrifuge tubes: For 15ml Tubes For 50ml Tubes Plastic boxes R8126 R8121 Cardboard boxes R3020 boxes + R1600-15-PK dividers R3020 boxes + R2000-16D-PK dividers Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our extensive product range will accommodate all type of sample containers including 2”, 3”, and 5” boxes, microtiter and deepwell plates and 15&50mL centrifuge tubes. Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright and Chest ultra low freezers.
Catalog No. 50-145-9967 Supplier Thomas Scientific Supplier No. 1158N59
Denville freezer boxes for 15ml and 50ml centrifuge tubes: For 15ml Tubes For 50ml Tubes Plastic boxes R8126 R8121 Cardboard boxes R3020 boxes + R1600-15-PK dividers R3020 boxes + R2000-16D-PK dividers Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our extensive product range will accommodate all type of sample containers including 2”, 3”, and 5” boxes, microtiter and deepwell plates and 15&50mL centrifuge tubes. Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright and Chest ultra low freezers.