Learn More
Thomas Scientific Rack Drawer for 9 2" Bx, 3x3
SDP
Catalog No. 501460898
Encompass
Argos PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our extensive product range will accommodate all type of sample containers including 2", 3", and 5" boxes, microtiter and deepwell plates and 15&50mL centrifuge tubes. Argos PolarSafe™ Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright and Chest ultra low freezers. Need help deciding which racks works best with your ultra low freezer? Try our Freezer Rack Configurator .
Catalog No. 50-146-0898 Supplier Thomas Scientific Supplier No. 1218D70
Argos PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our extensive product range will accommodate all type of sample containers including 2", 3", and 5" boxes, microtiter and deepwell plates and 15&50mL centrifuge tubes. Argos PolarSafe™ Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright and Chest ultra low freezers. Need help deciding which racks works best with your ultra low freezer? Try our Freezer Rack Configurator .