Learn More
Julabo USA Inc RACK FOR 21 TEST TUBES 30MM
SDP
Catalog No. 507534064
Encompass_Preferred
Stainless steel test tube rack for 21 tubes, 30/31 mm diameter. For use with JULABO shaking water baths SW22, SW23. Requires test tube rack carrier tray: 8970369.
Catalog No. 50-753-4064 Supplier Julabo USA Inc Supplier No. 8970347
Stainless steel test tube rack for 21 tubes, 30/31 mm diameter. For use with JULABO shaking water baths SW22, SW23. Requires test tube rack carrier tray: 8970369.