Learn More
MTC Bio Rack for 4 x 500mL centrifuge tubes, plastic coated wire, autoclavable, 1/ea
Catalog No. 502268452
Encompass_Preferred
500mL screw-cap polypropylene tubes are manufactured of polypropylene for temperature and chemical resistance. These tubes are intended for large volume centrifugation and are widely used in cell harvesting/purification and drug discovery applications. MTC Bio's 500mL centrifuge tubes can withstand up to 6,000xg maximum RCF*. They are supplied in sterile, RNase, DNase, and pyrogen-free packs of 5 tubes, 40 tubes per case.
This item is not returnable. View return policy
Catalog No. 50-226-8452 Supplier MTC Bio Supplier No. C1500RK
500mL screw-cap polypropylene tubes are manufactured of polypropylene for temperature and chemical resistance. These tubes are intended for large volume centrifugation and are widely used in cell harvesting/purification and drug discovery applications. MTC Bio's 500mL centrifuge tubes can withstand up to 6,000xg maximum RCF*. They are supplied in sterile, RNase, DNase, and pyrogen-free packs of 5 tubes, 40 tubes per case.