Learn More
Thomson Instrument Company 24 Position Block For 4mL Vials, Black
SDP
Catalog No. 50996050
Encompass_Preferred
Precision 24-well black metal block for 4 mL vials that adds heft and stability for dependable pipetting and sample handling. Ideal for method development benches where durable hardware is preferred. Made in the USA
Catalog No. 50-996-050 Supplier Thomson Instrument Company Supplier No. 1212205BK
Precision 24-well black metal block for 4 mL vials that adds heft and stability for dependable pipetting and sample handling. Ideal for method development benches where durable hardware is preferred. Made in the USA