Learn More
Tisch Scientific Tisch Scientific Vial Rack for 4 mL Vials, Blue Polypropylene, 9.25" x 4.5" x 1.125", 50 Holes, Hole Diameter: 15 mm, Hole Depth: 22 mm, Non-Autoclavable, 1 Pack, CV2056
SDP
Catalog No. NC1811804
Encompass
Tisch 4mL vial racks are designed for use in laboratories, medical settings, research and more. Vial racks are made from industry-standard polypropylene plastic and hold up to 50 vials per rack.
Catalog No. NC1811804 Supplier Tisch Scientific Supplier No. CV2056
Tisch 4mL vial racks are designed for use in laboratories, medical settings, research and more. Vial racks are made from industry-standard polypropylene plastic and hold up to 50 vials per rack.