Learn More
Julabo USA Inc Each, test tube rack, 50 tubes 17mm diameter, stainless steel
SDP
Catalog No. 507534061
Encompass_Preferred
Bath Accessories, Julabo, Test tube rack, for 50 test tubes, 15/16 or 16/17 mm, diameter, to +100°C. For use with shaking water baths SW22, SW23. Requires base tray 8970369. Stainless steel.
Catalog No. 50-753-4061 Supplier Julabo USA Inc Supplier No. 8970344
Bath Accessories, Julabo, Test tube rack, for 50 test tubes, 15/16 or 16/17 mm, diameter, to +100°C. For use with shaking water baths SW22, SW23. Requires base tray 8970369. Stainless steel.