Learn More
MTC Bio SureStand™ Pipette Stand for 6 Halo™ Single Channel Pipettes or 6 ProPette LE Single Channel Pipettes, up to four ProPette LE Multi-channel Pipettes
Catalog No. NC2017370
Encompass_Preferred
SureStand™ Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patent pending “windows” allow the user to insert the multi-channel head long ways through the rack, while keeping the body of the pipette stable by securing the pipette at two points; under the finger hook, and near the bottom of the handle. SureStand is available in 3 versions; a 3-place rack that accepts one multi-channel, a 5-place rack that accepts two multi-channels, and a 6-place rack that accepts four multichannels. Each SureStand is also outfitted with four non-skid rubber “feet” for extra stability. SureStand is guaranteed to be the sturdiest and most versatile pipette rack in your lab
Catalog No. NC2017370 Supplier MTC Bio Supplier No. P4406P
SureStand™ Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patent pending “windows” allow the user to insert the multi-channel head long ways through the rack, while keeping the body of the pipette stable by securing the pipette at two points; under the finger hook, and near the bottom of the handle. SureStand is available in 3 versions; a 3-place rack that accepts one multi-channel, a 5-place rack that accepts two multi-channels, and a 6-place rack that accepts four multichannels. Each SureStand is also outfitted with four non-skid rubber “feet” for extra stability. SureStand is guaranteed to be the sturdiest and most versatile pipette rack in your lab