Learn More
MTC Bio Ice Rack™ for 72 × 0.5mL tubes, detachable lid, tube platform, and base, numbered cavities, accepts snap cap and screw cap 0.5mL to 0.65mL tubes, 6/cs
Catalog No. 502268489
Encompass_Preferred
These new tube boxes have a removable white tube tray that can be kept filled with crushed ice or other temperature controlling materials. They also have a translucent cover. Temperature sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. Suitable for use with screw cap or snap cap microtubes from 0.5mL to 1.5/2.0mL Supplied 6 boxes per pack.
This item is not returnable. View return policy
Catalog No. 50-226-8489 Supplier MTC Bio Supplier No. R8012
These new tube boxes have a removable white tube tray that can be kept filled with crushed ice or other temperature controlling materials. They also have a translucent cover. Temperature sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. Suitable for use with screw cap or snap cap microtubes from 0.5mL to 1.5/2.0mL Supplied 6 boxes per pack.