Learn More
Julabo USA Inc RACK FOR 90 UL TUBES 11/12MM
SDP
Catalog No. 507534063
Encompass_Preferred
Stainless steel test tube rack for 90 tubes, 11/12 mm diameter. For use with JULABO shaking water baths SW22, SW23. Requires test tube rack carrier tray: 8970369.
Catalog No. 50-753-4063 Supplier Julabo USA Inc Supplier No. 8970346
Stainless steel test tube rack for 90 tubes, 11/12 mm diameter. For use with JULABO shaking water baths SW22, SW23. Requires test tube rack carrier tray: 8970369.