Learn More
Microsolv Technology Corporation Rack for Headspace Vials, Clear acrylic plastic. 25 cavities with a diameter of 24mm. 1 EA.
Catalog No. 502271218
Encompass_Preferred
Often used for storage, transportation or filling, this heavy weight rack is excellent for 20mm wide Headspace Vials. The clear acrylic makes is possible to see into the bottom of the vials for problems with your samples.Dimensions: 160mm x 160mm x 30mm.
This item is not returnable. View return policy
Catalog No. 50-227-1218 Supplier Microsolv Technology Corporation Supplier No. 95025R
Often used for storage, transportation or filling, this heavy weight rack is excellent for 20mm wide Headspace Vials. The clear acrylic makes is possible to see into the bottom of the vials for problems with your samples.Dimensions: 160mm x 160mm x 30mm.