Learn More
Thomas Scientific Rack for Microcentrifuge Tubes, 96 positions; polypropylene, blue, 5 pack
SDP
Catalog No. NC1682291
Encompass
Polypropylene (PP), reversible 96-well rack in an 8 x 12 format, one side holds 1.5 mL, 1.8 mL, or 2.0 mL microcentrifuge tubes; the other side holds 0.5 mL tubes, and alpha-numerically labeled, stackable, autoclavable.
Catalog No. NC1682291 Supplier Thomas Scientific Supplier No. 1178M76
Polypropylene (PP), reversible 96-well rack in an 8 x 12 format, one side holds 1.5 mL, 1.8 mL, or 2.0 mL microcentrifuge tubes; the other side holds 0.5 mL tubes, and alpha-numerically labeled, stackable, autoclavable.