Learn More
Sigma Aldrich Fine Chemicals Biosciences Rack for NalgeneR microcen
Catalog No. 502900101
Encompass
Bottom of white polycarbonate lid of clear polycarbonate permanent foam insert.for safe storage of samples in microcentrifuge tubes from -135AC to 121AC. Not for use in liquid-phase liquid nitrogen. May be repeatedly autoclaved. Holes in foam are placed diagonally and spaced to avoid overlapping of tube closures. Lid accepts writing with marker.
Catalog No. 50-290-0101 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R12634EA
Add to Cart
Add to Cart
Bottom of white polycarbonate lid of clear polycarbonate permanent foam insert.for safe storage of samples in microcentrifuge tubes from -135AC to 121AC. Not for use in liquid-phase liquid nitrogen. May be repeatedly autoclaved. Holes in foam are placed diagonally and spaced to avoid overlapping of tube closures. Lid accepts writing with marker.