Learn More
Research Products International Corp Upright Freezer Rack for 50 and 15 ml Tubes, 3 x 2 Array, 5 7/8 x 9 7/8 x 18 1/4 Inch
SDP
Catalog No. 50550277
Encompass
Maximize your freezer storage with a freezer rack. The freezer rack can fit 5 3/4 inch square boxes with a 4 3/4 inch height. Box capacity is 6 in a 3 x 2 array. Boxes measuring 5 3/4 x 4 3/4 inch boxes are for storage of 15 and 50 ml tubes.
Catalog No. 50-550-277 Supplier Research Products International Corp Supplier No. 247580
Add to Cart
Add to Cart
Maximize your freezer storage with a freezer rack. The freezer rack can fit 5 3/4 inch square boxes with a 4 3/4 inch height. Box capacity is 6 in a 3 x 2 array. Boxes measuring 5 3/4 x 4 3/4 inch boxes are for storage of 15 and 50 ml tubes.