Learn More
MTC Bio Magnetic bead separation rack, reversible, for 8x1.5mL Tube or 16x0.2mL tubes
Catalog No. NC3737137
Encompass_Preferred
MTC Bio’s reversible magnetic rack is ideal for low throughput bead-based nucleic acid purification. Double-sided magnets combined with an innovative flip-design allows for bead separation in 8 x 1.5/2.0mL microtubes, or 16 x 0.2mL PCR tubes, all while maintaining an exceptionally compact footprint. The ultra-strong magnets feature a Pull Strength of 5000g and Field Strength of more than 5000 guass for fast separation. Tube wells are angled on a 10° pitch for optimal tube contact with magnets, and microtube wells are equipped with an adapter ring for precision control over the height of the tube exposed to the magnetic field. The magnets are housed in a removable casing that enables quick resuspension without removing tubes from the rack.• Flip-over design for Microtubes or PCR Tubes• Powerful magnets for quick and efficient separation • Removable magnet housing for resuspension • Adapter rings for variable tube height• Ultra-comp
Catalog No. NC3737137 Supplier MTC Bio Supplier No. R0215
MTC Bio’s reversible magnetic rack is ideal for low throughput bead-based nucleic acid purification. Double-sided magnets combined with an innovative flip-design allows for bead separation in 8 x 1.5/2.0mL microtubes, or 16 x 0.2mL PCR tubes, all while maintaining an exceptionally compact footprint. The ultra-strong magnets feature a Pull Strength of 5000g and Field Strength of more than 5000 guass for fast separation. Tube wells are angled on a 10° pitch for optimal tube contact with magnets, and microtube wells are equipped with an adapter ring for precision control over the height of the tube exposed to the magnetic field. The magnets are housed in a removable casing that enables quick resuspension without removing tubes from the rack.• Flip-over design for Microtubes or PCR Tubes• Powerful magnets for quick and efficient separation • Removable magnet housing for resuspension • Adapter rings for variable tube height• Ultra-comp