Learn More
Chemglass Life Sciences Rack, Reversible, Red, Polypropylene, Micro Tube
SDP
Catalog No. 501532374
Encompass_Preferred
Rack, Reversible, Red, Polypropylene, Micro Tube. Reversible polypropylene micro tube racks with secure grip handles. Side 1 holds (80) 1.5/2.0mL micro tubes, Side 2 holds (60) 0.5mL micro tubes. Ideal for benchtop or freezer use. Autoclavable. Dimensions (mm): 228 L x 67 W x 27 H.
Catalog No. 50-153-2374 Supplier Chemglass Life Sciences Supplier No. CLS477705R
Rack, Reversible, Red, Polypropylene, Micro Tube. Reversible polypropylene micro tube racks with secure grip handles. Side 1 holds (80) 1.5/2.0mL micro tubes, Side 2 holds (60) 0.5mL micro tubes. Ideal for benchtop or freezer use. Autoclavable. Dimensions (mm): 228 L x 67 W x 27 H.