Learn More
Electron Microscopy Sciences Microwaveable Square Micro Tube Rack, White
SDP
Catalog No. 5034079
Encompass_Preferred
Polypropylene floating racks keep microtubes submerged in a water bath – perfect for controlling sample temperature. Detachable legs give the users more options: attach legs so rack stands upright on benchtop, or detach legs so sample tubes can be quickly popped out by pushing down onto any surface. Ideally to hold microcentrifuge tube in water bath during polymerization, such as LR White, JB4. Not autoclavable. Measures: 60 mm high, including legs.
Color: White
Catalog No. 50-340-79 Supplier Electron Microscopy Sciences Supplier No. 72372SB
Polypropylene floating racks keep microtubes submerged in a water bath – perfect for controlling sample temperature. Detachable legs give the users more options: attach legs so rack stands upright on benchtop, or detach legs so sample tubes can be quickly popped out by pushing down onto any surface. Ideally to hold microcentrifuge tube in water bath during polymerization, such as LR White, JB4. Not autoclavable. Measures: 60 mm high, including legs.
Color: White