Learn More
Alkali Scientific PCR Test Tube Rack, 96 x 0.2 ml, Raimbow Pack of 5
SDP
Catalog No. 502336184
Encompass_Preferred
Suitable for refrigerator and freezer storage as low as -80°C, these boxes have translucent lids and numbered cavities. They are a PCR workstation rack/box for 96 x 0.2ml (plates, strips or individual tubes).
Catalog No. 50-233-6184 Supplier Alkali Scientific Supplier No. AR1010
Suitable for refrigerator and freezer storage as low as -80°C, these boxes have translucent lids and numbered cavities. They are a PCR workstation rack/box for 96 x 0.2ml (plates, strips or individual tubes).