Learn More
Hach Company Pipet Rack
Catalog No. 501995353 Shop All Hach Company Products
Encompass
Polypropylene, fifty-place rack for safe, orderly storage of pipets. Helps create an efficient storage system or a well organized liquid handling station. Holes in top and middle decks are 16 mm diameter (0.63"); bottom holes are tapered to 6 mm (0.24") to prevent contact with supporting surface. Autoclavable to 122°C. Overall dimension: 21 x 11 x 22 cm (8.3 x 4.3 x 8.7").
Catalog No. 50-199-5353 Supplier Hach Company Supplier No. 2094700
Polypropylene, fifty-place rack for safe, orderly storage of pipets. Helps create an efficient storage system or a well organized liquid handling station. Holes in top and middle decks are 16 mm diameter (0.63"); bottom holes are tapered to 6 mm (0.24") to prevent contact with supporting surface. Autoclavable to 122°C. Overall dimension: 21 x 11 x 22 cm (8.3 x 4.3 x 8.7").