Learn More
Cole-Parmer Argos Technologies Acrylic Pipette Organizing Rack; Clear
Supplier:  Cole-Parmer 2440704
Encompass_Preferred
Heavy-duty, acrylic rack features five angled compartments which can house 1 to 100 mL glass or plastic pipettes. Two slots at the top of the rack are designed to hold pipette controllers. Available in blue-tinted or clear acrylic colors.
Catalog No. 50-196-0868