Learn More
Thomas Scientific Pipet Storage Rack, Plastic, Magnetized
SDP
Catalog No. 501457751
Encompass_Preferred
Durable, ABS plastic rack can stand on flat surfaces as well as be attached to the metal sides of lab cabinets and appliances. Strong magnets are included on both sides of the rack for increased versatility. Strong corner magnets for attachment to metal fume hoods, refrigerators and cabinets Angled shelves provide secure yet easy access to pipets Pipet sizes, colors and types can be viewed from the side windows as well as the front Side bracket is handy for holding pipet fillers, bulbs and pumps
Catalog No. 50-145-7751 Supplier Thomas Scientific Supplier No. 1195W22
Durable, ABS plastic rack can stand on flat surfaces as well as be attached to the metal sides of lab cabinets and appliances. Strong magnets are included on both sides of the rack for increased versatility. Strong corner magnets for attachment to metal fume hoods, refrigerators and cabinets Angled shelves provide secure yet easy access to pipets Pipet sizes, colors and types can be viewed from the side windows as well as the front Side bracket is handy for holding pipet fillers, bulbs and pumps