Learn More
Bio Rad Laboratories RACK SET F3
Catalog No. 509010338
Encompass
Set F3, Rack; Bio-Rad; Accessory for BioFrac Fraction Collector; 2 x flatpack; Each holds 40 tubes, 18-20mm diameter, for total of up to 80 tube collection
Catalog No. 50-901-0338 Supplier Bio Rad Laboratories Supplier No. 7410012
Set F3, Rack; Bio-Rad; Accessory for BioFrac Fraction Collector; 2 x flatpack; Each holds 40 tubes, 18-20mm diameter, for total of up to 80 tube collection