Learn More
Bio Rad Laboratories RACK SET H4
Catalog No. 509010326
Encompass
Set H4, Rack; Bio-Rad; Accessory for BioFrac Fraction Collector; 4 x flatpack; Each holds 6 scintillation vials, 30mm diameter, for total of 24-vial collection
Catalog No. 50-901-0326 Supplier Bio Rad Laboratories Supplier No. 7410016
Set H4, Rack; Bio-Rad; Accessory for BioFrac Fraction Collector; 4 x flatpack; Each holds 6 scintillation vials, 30mm diameter, for total of 24-vial collection