Learn More
Andwin Scientific RACK SYSTEM MODULAR PP EA
SDP
Catalog No. 501305881
Encompass_Preferred
This modular polypropylene test tube rack system is composed of one support tray and four color-coded cube modules that can be rotated independently to be used for several sizes of test tubes, culture tubes or microcentrifuge tubes at the same time. It is compatible with tubes such as: 15- and 50-mL conical; 12 x 75 mm, 13 x 100 mm, 16 x 125 mm, and 17 x 100 mm culture; and 1.5- and 2.0-mL.
Catalog No. 50-130-5881 Supplier Andwin Scientific Supplier No. 673300A
This modular polypropylene test tube rack system is composed of one support tray and four color-coded cube modules that can be rotated independently to be used for several sizes of test tubes, culture tubes or microcentrifuge tubes at the same time. It is compatible with tubes such as: 15- and 50-mL conical; 12 x 75 mm, 13 x 100 mm, 16 x 125 mm, and 17 x 100 mm culture; and 1.5- and 2.0-mL.