Learn More
Cole-Parmer Autoclavable PP Modular Test Tube Rack System
Catalog No. NC9475210 Shop All Cole-Parmer Products
Encompass_Preferred
Cube modules are color coded for convenience. Rotate each cube independently—use several sizes of test tubes, culture tubes, or microcentrifuge tubes at the same time. Each system consists of four cubes and one support tray. Compatible with the following tubes: 15- and 50-mL conical; 12 x 75 mm, 13 x 100 mm, 16 x 125 mm, and 17 x 100 mm culture; and 1.5- and 2.0-mL microcentrifuge. Autoclavable.
Catalog No. NC9475210 Supplier Cole-Parmer Supplier No. 0673300
Cube modules are color coded for convenience. Rotate each cube independently—use several sizes of test tubes, culture tubes, or microcentrifuge tubes at the same time. Each system consists of four cubes and one support tray. Compatible with the following tubes: 15- and 50-mL conical; 12 x 75 mm, 13 x 100 mm, 16 x 125 mm, and 17 x 100 mm culture; and 1.5- and 2.0-mL microcentrifuge. Autoclavable.