Learn More
Hach Company Rack, Test Tube, Polyethylene, 13 mm (O.D.), 90 Holes
Supplier:  Hach Company 2497900
Encompass
Twenty to ninety-place rack features a special blend of polypropylene which sinks in water, assuring non-floating racks when immersed. Autoclavable. Stackable and autoclavable. Overall dimensions: 24.6 x 10.7 x 6.4 cm (9.7 x 4.2 x 2.5").
Catalog No. 50-199-6087