Learn More
Waters Corp Test Tube Rack, 20-Port, 16 x 100 mm
Catalog No. 502392349 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This test tube rack is designed for 16 x 100 mm test tubes and is compatible with a wide range of laboratory equipment. It provides a stable area to hold samples for analysis, making it a vital component in various laboratory processes.
  • Compatible with vacuum manifold for use with SPE cartridges.
  • Can process up to 20 samples simultaneously.
  • Designed for traditional silica-based SPE cartridges.
  • Modified to utilize high-performance extraction cartridges.
  • Provides a stable area to hold samples for analysis.
Catalog No. 50-239-2349 Supplier Waters Corp Supplier No. 186008997
This test tube rack is designed for 16 x 100 mm test tubes and is compatible with a wide range of laboratory equipment. It provides a stable area to hold samples for analysis, making it a vital component in various laboratory processes.
  • Compatible with vacuum manifold for use with SPE cartridges.
  • Can process up to 20 samples simultaneously.
  • Designed for traditional silica-based SPE cartridges.
  • Modified to utilize high-performance extraction cartridges.
  • Provides a stable area to hold samples for analysis.