Learn More
Hach Company Test Tube/ MPN Rack, Polyethylene, 20 mm (O.D.), 40 Holes
Catalog No. 501996089 Shop All Hach Company Products
Encompass
Twenty to ninety-place rack features a special blend of polypropylene which sinks in water, assuring non-floating racks when immersed. Autoclavable. Stackable and autoclavable. Overall dimensions: 24.6 x 10.7 x 6.4 cm (9.7 x 4.2 x 2.5").
Catalog No. 50-199-6089 Supplier Hach Company Supplier No. 2497902
Twenty to ninety-place rack features a special blend of polypropylene which sinks in water, assuring non-floating racks when immersed. Autoclavable. Stackable and autoclavable. Overall dimensions: 24.6 x 10.7 x 6.4 cm (9.7 x 4.2 x 2.5").