Learn More
Hach Company Rack, Test Tube, Polyethylene
Catalog No. 501995145 Shop All Hach Company Products
Encompass
Twelve-place rack accepts up to 24 mm (O.D.) tubes and has five pins for inverting tubes for drying. Autoclavable.
Catalog No. 50-199-5145 Supplier Hach Company Supplier No. 1488600
Twelve-place rack accepts up to 24 mm (O.D.) tubes and has five pins for inverting tubes for drying. Autoclavable.