Learn More
Hach Company Rack-Test Tube-Polypropylene
Supplier:  Hach Company 2407000
Encompass
Test tube rack made of Polypropylene Will not float in water and autoclavable Molded in one piece stacks easily Tube size 20 mm (0 86in ) array 4 x 10 Dimensions 25 x 10 2 x 8 3 cm (9 75 x 4 x 3 25in ) Nalge
Catalog No. 50-002-57132