Learn More
Labcon North America Refillable Racks for Eclipse™ 100-1250uL Refill Tips, Autoclavable
Catalog No. 50101721
Encompass_Preferred
Rack; Pipet tip; Refillable; With hinged clear lid; For use with Eclipse™ pipet tip refills; For use with 100-1250uL tips; Holds one refill platform of 96 tips; Autoclavable; Teal color base; 6/pk; 8/pk per case; Made in USA
Catalog No. 50-101-721 Supplier Labcon North America Supplier No. 12799600089
Rack; Pipet tip; Refillable; With hinged clear lid; For use with Eclipse™ pipet tip refills; For use with 100-1250uL tips; Holds one refill platform of 96 tips; Autoclavable; Teal color base; 6/pk; 8/pk per case; Made in USA