Learn More
Beckman Coulter Microplate Rack, 96-Well, Polypropylene, Clear
Catalog No. NC1157901
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge Rotor is a fixed-angle rotor designed for high-speed microcentrifugation. It has a maximum speed of 18,000 rpm and maximum RCF of 30,130 xg. The rotor has a 6-place capacity for 1.5/2.0mL microcentrifuge tubes. It is constructed of durable polycarbonate and has an aluminum hub and cover for thermal conductivity. The rotor is compatible with multiple Beckman Coulter microcentrifuges including the Microfuge 18, Microfuge 20, and Microfuge 22 series. It provides fast and efficient microcentrifugation for a variety of laboratory applications.
Catalog No. NC1157901 Supplier Beckman Coulter Supplier No. 358991
The Beckman Coulter Microfuge 18 Centrifuge Rotor is a fixed-angle rotor designed for high-speed microcentrifugation. It has a maximum speed of 18,000 rpm and maximum RCF of 30,130 xg. The rotor has a 6-place capacity for 1.5/2.0mL microcentrifuge tubes. It is constructed of durable polycarbonate and has an aluminum hub and cover for thermal conductivity. The rotor is compatible with multiple Beckman Coulter microcentrifuges including the Microfuge 18, Microfuge 20, and Microfuge 22 series. It provides fast and efficient microcentrifugation for a variety of laboratory applications.