Learn More
Med Vet International McKesson Tube Cube Rack, 3 x 3 x 3 Inch, 3096, 4/Box
Catalog No. 503045773
Encompass_Preferred
Tube Cube Rack McKesson 9 Place 8 to 16 mm Tube Size Blue 3 X 3 X 3 Inch Test tube rack holds nine 8 – 16 mm tubes 3 x 3 x 3 inch test tube rack not only holds tubes but also cradles their bottoms; the middle section also grasps the tubes at their mid point: Your test tubes will definitely be secure in this rack Test tube rack is intended for laboratory use but those who breed monarch butterflies also find them useful to set up milkweed cuttings
Catalog No. 50-304-5773 Supplier Med Vet International Supplier No. 885932BX
Tube Cube Rack McKesson 9 Place 8 to 16 mm Tube Size Blue 3 X 3 X 3 Inch Test tube rack holds nine 8 – 16 mm tubes 3 x 3 x 3 inch test tube rack not only holds tubes but also cradles their bottoms; the middle section also grasps the tubes at their mid point: Your test tubes will definitely be secure in this rack Test tube rack is intended for laboratory use but those who breed monarch butterflies also find them useful to set up milkweed cuttings