Learn More
USA Scientific Inc 10 ul XL TipOne filter tip, sterile, case of 5 packs
Catalog No. 50143955
Encompass
TipOne 10 ul XL aerosol barrier filter pipet tips, sterile, 50 racks of 96 tips (4800 tips)
Catalog No. 50-143-955 Supplier USA Scientific Inc Supplier No. 11203860
TipOne 10 ul XL aerosol barrier filter pipet tips, sterile, 50 racks of 96 tips (4800 tips)