Learn More
Chemglass Life Sciences Freezer Rack with Two Drawers, Upright Drawer Style, for Centrifuge Tubes, Holds (160) 15mL Tubes, 22"L x 5-1/2"W x 9-15/16"H
SDP
Catalog No. 50143315
Encompass
Freezer Rack with Two Drawers, Upright Drawer Style, for Centrifuge Tubes, Holds (160) 15mL Tubes, 22"L x 5-1/2"W x 9-15/16"H. Drawers make these freezer racks easy and convenient to use. No need to remove the entire rack from the freezer to gain access, just slide the drawer out.Centrifuge tube racks are specifically designed to provide an alternative for storing large quantities of samples in 15mL and 50mL centrifuge tubes. Racks for two-inch boxes hold between nine and twenty-four boxes.High quality racks are constructed of corrosion-resistant stainless steel and include handles and identification card holders. Secure lock devices are available ¿ please contact us for more information.
Catalog No. 50-143-315 Supplier Chemglass Life Sciences Supplier No. CLS3795101
Freezer Rack with Two Drawers, Upright Drawer Style, for Centrifuge Tubes, Holds (160) 15mL Tubes, 22"L x 5-1/2"W x 9-15/16"H. Drawers make these freezer racks easy and convenient to use. No need to remove the entire rack from the freezer to gain access, just slide the drawer out.Centrifuge tube racks are specifically designed to provide an alternative for storing large quantities of samples in 15mL and 50mL centrifuge tubes. Racks for two-inch boxes hold between nine and twenty-four boxes.High quality racks are constructed of corrosion-resistant stainless steel and include handles and identification card holders. Secure lock devices are available ¿ please contact us for more information.