Learn More
Chemglass Life Sciences Racks, Upright Style Freezer, for 12 (4L x 3H) 3" Boxes, 21-5/8"L x 5-1/2"W x 9-3/8"H
SDP
Catalog No. 50143304
Encompass
Upright Freezer Drawer Racks are for standard 3¿ cardboard, plastic (polycarbonate) or metal (stainless steel or aluminum) boxes. High quality upright freezer racks are constructed of corrosion resistant stainless steel, including handles and identification card holders. Racks offer a more organized system for storage and retrieval of samples. They facilitate both access to and storage of samples without having to remove the whole rack from the freezer.
Catalog No. 50-143-304 Supplier Chemglass Life Sciences Supplier No. CLS3792301
Upright Freezer Drawer Racks are for standard 3¿ cardboard, plastic (polycarbonate) or metal (stainless steel or aluminum) boxes. High quality upright freezer racks are constructed of corrosion resistant stainless steel, including handles and identification card holders. Racks offer a more organized system for storage and retrieval of samples. They facilitate both access to and storage of samples without having to remove the whole rack from the freezer.