Learn More
Chemglass Life Sciences Racks, Vertical Style Freezer, for (10) 2" Boxes, 5-5/8"L x 5-1/2"W x 22-1/16"H
SDP
Catalog No. 50143299
Encompass
Racks, Vertical Style Freezer, for (10) 2" Boxes, 5-5/8"L x 5-1/2"W x 22-1/16"H. High quality vertical freezer racks for chest freezers or liquid nitrogen tanks are made from corrosion resistant stainless steel. Secure lock devices are available - please contact us for details. All racks include stainless steel locking rods.
Catalog No. 50-143-299 Supplier Chemglass Life Sciences Supplier No. CLS3793001
Racks, Vertical Style Freezer, for (10) 2" Boxes, 5-5/8"L x 5-1/2"W x 22-1/16"H. High quality vertical freezer racks for chest freezers or liquid nitrogen tanks are made from corrosion resistant stainless steel. Secure lock devices are available - please contact us for details. All racks include stainless steel locking rods.