Learn More
Chemglass Life Sciences Racks, Vertical Style Freezer, for (12) 2" Boxes, 5-5/8"L x 5-1/2"W x 26-3/8"H
SDP
Catalog No. 50143451
Encompass
Racks, Vertical Style Freezer, for (12) 2" Boxes, 5-5/8"L x 5-1/2"W x 26-3/8"H. High quality vertical freezer racks for chest freezers or liquid nitrogen tanks are made from corrosion resistant stainless steel. Secure lock devices are available - please contact us for details. All racks include stainless steel locking rods.
Catalog No. 50-143-451 Supplier Chemglass Life Sciences Supplier No. CLS3793003
Racks, Vertical Style Freezer, for (12) 2" Boxes, 5-5/8"L x 5-1/2"W x 26-3/8"H. High quality vertical freezer racks for chest freezers or liquid nitrogen tanks are made from corrosion resistant stainless steel. Secure lock devices are available - please contact us for details. All racks include stainless steel locking rods.