Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
Catalog No. 13874770
Change view
Click to view available options
Type:
Rack
Rack Insert
Holds:
100-10 mm Tubes
13-25 mm Tubes
20-16 mm Tubes
32-16 mm Tubes
39-10 mm Tubes
60-16 mm Tubes
Depends on insert purchased
Dishes, plates, bags, pouches
For Use With (Equipment):
A25B, A40, S21, S30
A5B,A10B A24B,S49,S19T,S14P,S21P
S13, S12T, S15
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Holds For Use With (Equipment)
13-874-770 Rack Depends on insert purchased A5B,A10B A24B,S49,S19T,S14P,S21P
13-255-80 Rack Insert 100-10 mm Tubes A5B,A10B A24B,S49,S19T,S14P,S21P
13-255-60 Rack Insert 60-16 mm Tubes A5B,A10B A24B,S49,S19T,S14P,S21P
13-255-63 Rack Depends on insert purchased A25B, A40, S21, S30
13-255-65 Rack Insert 32-16 mm Tubes A25B, A40, S21, S30
13-255-651 Rack Insert 13-25 mm Tubes A25B, A40, S21, S30
13-255-652 Rack Insert Dishes, plates, bags, pouches A25B, A40, S21, S30
13-255-654 Rack Insert 39-10 mm Tubes S13, S12T, S15
13-255-655 Rack Insert 20-16 mm Tubes S13, S12T, S15
13-872-270 Rack Insert Dishes, plates, bags, pouches S13, S12T, S15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 13-874-770 Supplier Thermo Scientific™ Supplier No. 1600002
Add to Cart
Add to Cart
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) A5B,A10B A24B,S49,S19T,S14P,S21P
Type Rack
Holds Depends on insert purchased
Material Stainless Steel