Learn More
USA Scientific Inc 9-place (3 x 3) upright freezer sliding rack for 2 inch H boxes
Catalog No. NC9638337
Encompass
9-place (3 long x 3 high) upright freezer sliding rack for 2 in H boxes, 16 1/2 in L x 5 1/2 in W x 7 1/8 in H, stainless steel
Catalog No. NC9638337 Supplier USA Scientific Inc Supplier No. 26323300
9-place (3 long x 3 high) upright freezer sliding rack for 2 in H boxes, 16 1/2 in L x 5 1/2 in W x 7 1/8 in H, stainless steel