Learn More
Chemglass Life Sciences Racks, Upright Style Freezer, for 96 and 384-Well Microtiter Plates, Holds 96 With Lids / 120 Without Lids, 22"L x 5-1/2"W x 11"H
SDP
Catalog No. 50143443
Encompass
Racks, Upright Style Freezer, for 96 and 384-Well Microtiter Plates, Holds 96 With Lids / 120 Without Lids, 22"L x 5-1/2"W x 11"H. High quality freezer racks for 96-well and 384-well microtiter plates, including handles, constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format. Security lock devices are also available for each rack. Racks come with locking rods to secure microtiter plates in place.
Catalog No. 50-143-443 Supplier Chemglass Life Sciences Supplier No. CLS3794005
Racks, Upright Style Freezer, for 96 and 384-Well Microtiter Plates, Holds 96 With Lids / 120 Without Lids, 22"L x 5-1/2"W x 11"H. High quality freezer racks for 96-well and 384-well microtiter plates, including handles, constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format. Security lock devices are also available for each rack. Racks come with locking rods to secure microtiter plates in place.